missing translation for 'onlineSavingsMsg'
Learn More

Clanobutin, TRC

Product Code. 30132925
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132925 100 mg 100mg
30004875 10 mg 10mg
30006240 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132925 Supplier Toronto Research Chemicals Supplier No. TRCC558000100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30544-61-7
Molecular Formula C18 H18 Cl N O4
Molecular Weight (g/mol) 347.79
Synonym Clanobutin, 4-[(4-Chlorobenzoyl)(4-methoxyphenyl)amino]butanoic acid (ACI), Butyric acid, 4-[p-chloro-N-(p-methoxyphenyl)benzamido]- (8CI), Bykahepar, Clanobutin, Clanobutine, N-(p-Chlorobenzoyl)-γ-(p-anisidino)butyric acid
IUPAC Name 4-(N-(4-chlorobenzoyl)-4-methoxyanilino)butanoic acid
SMILES COc1ccc(cc1)N(CCCC(=O)O)C(=O)c2ccc(Cl)cc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H18ClNO4/c1-24-16-10-8-15(9-11-16)20(12-2-3-17(21)22)18(23)13-4-6-14(19)7-5-13/h4-11H,2-3,12H2,1H3,(H,21,22)
Formula Weight 347.0924
Percent Purity >95%
Chemical Name or Material Clanobutin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.