Learn More
Clamps
Flasks clamps for shaking incubators
Brand: Medline Scientific AAA23551
Code : 04
Additional Details : Weight : 0.50000kg
Description
The Lab Companion range of refrigerated shaking incubators include the SI-600R, a smaller version, the SI-300R and the floor model IS-971R which is a capable of incubating large volumes and very low temperatures (+4°C). The SI-300R and SI-600R units are bench top models with a slim width to ensure they fit into the tightest of spaces. The lid, which is supported by integral gas pressure struts, opens to give front and top access for any size flask up to 2, 000mL. The patented CLS control system exceeds Class II safety standards for use in areas where there may be flammable environments.- PID controller ensures accurate temperature control
- LED display
- Select orbital or reciprocal motion
- Cooling coils supplied with non-refrigerated models
- Adjustable stroke length
- Supplied with universal platform Range of bench top and floorstanding shaking incubators suitable for a variety of applications.
Specifications
100 mL Round Flask | |
Clamp |