Learn More
DWK Life Sciences Clamp for Spherical Joints, with tightening screw, made of stainless steel
Ball joint clamp with additional locking option.
Brand:  DWK Life Sciences 286335001
Additional Details : Weight : 1.25000kg
 View more versions of this product
Product Code. 15864678
  • £251.00 / Pack of 5
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
  • The locking screw enables a very secure connection by means of an additional locking device.
  • Excellent dimensional stability compared to conventional wire clamps
Specifications
Specifications
5
Securing of Spherical Ground Joints
S 64
Stainless Steel
Securing of spherical ground joints