Learn More
DWK Life Sciences Clamp for Spherical Joints, with tightening screw, made of stainless steel
Ball joint clamp with additional locking option.
Brand: DWK Life Sciences 286333806
Code : NEW
Additional Details : Weight : 0.05900kg
Description
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
- The locking screw enables a very secure connection by means of an additional locking device.
- Excellent dimensional stability compared to conventional wire clamps
Specifications
50 | |
Securing of Spherical Ground Joints | |
S 29 |
Stainless Steel | |
Securing of spherical ground joints |