Learn More
DWK Life Sciences™ Clamp for Spherical Joints, with tightening screw, made of stainless steel
Product Code. 10141781 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
S 13
S 19
S 29
S 35
S 40 and S 41
S 51
S 64
Unit Size:
Each
Pack of 10
Pack of 25
Pack of 5
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10141781 S 19 Each
10623982 S 35 Each
15854678 S 51 Pack of 10
15844678 S 40 and S 41 Pack of 25
10409342 S 29 Each
15864678 S 64 Pack of 5
15921571 S 13 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10141781 Supplier DWK Life Sciences™ Supplier No. 286333403
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ball joint clamp with additional locking option.
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
  • The locking screw enables a very secure connection by means of an additional locking device.
  • Excellent dimensional stability compared to conventional wire clamps
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50
Material Stainless Steel
For Use With (Equipment) Securing of Spherical Ground Joints
For Use With (Application) Securing of spherical ground joints
Size S 19