missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CL-P1/COLEC12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Applications | Western Blot |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
CL-P1/COLEC12 Polyclonal specifically detects CL-P1/COLEC12 in Human samples. It is validated for Western Blot.Specifications
| Western Blot | |
| Unconjugated | |
| RUO | |
| CL-P1, CLP1hCL-P1, collectin placenta 1, Collectin placenta protein 1, collectin sub-family member 12, collectin-12, Nurse cell scavenger receptor 2, SCARA4NSR2, Scavenger receptor class A member 4, scavenger receptor class A, member 4, SRCLScavenger receptor with C-type lectin | |
| COLEC12 | |
| IgG |
| Polyclonal | |
| Rabbit | |
| Q5KU26 | |
| 81035 | |
| Synthetic peptides corresponding to COLEC12(collectin sub-family member 12) The peptide sequence was selected from the N terminal of COLEC12. Peptide sequence AISTNSELSTFRSDILDLRQQLREITEKTSKNKDTLEKLQASGDALVDRQ. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title