missing translation for 'onlineSavingsMsg'
Learn More

CL-K1/COLEC11 Antibody (148), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18371031 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371031 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371031 Supplier Novus Biologicals Supplier No. NBP290474AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CL-K1/COLEC11 Monoclonal specifically detects CL-K1/COLEC11 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CL-K1/COLEC11
Applications ELISA
Classification Monoclonal
Clone 148
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias CLK1, CL-K1, CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, collectin kidney I, Collectin kidney protein 1, collectin sub-family member 11, collectin-11, DKFZp686N1868, MGC3279
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CL-K1/COLEC11 (Q3SXB8-1; Met1-Leu272).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78989
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.