missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CL 316243 disodium salt

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and highly selective β3-adrenoceptor agonist (EC50 = 3 nM); > 10000-fold selective over β1 and β2 receptors. Increases brown adipose tissue thermogenesis and metabolic rate, and decreases blood insulin and glucose levels following oral administration in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 465.8 |
| Purity | >96% |
| Molecular Weight (g/mol) | 442.804 |
| Chemical Name or Material | CL 316243 disodium salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction