Learn More
Olympus™ CKX3-SLP Phase Contrast Slider, Pre-centered for CKX53 and IX53
Click to view available options
For Use With (Equipment):
CKX53 and IX53 Microscopes
Unit Size:
Each
Description
- Converts phase shifts in light.
- Separates illuminating, background light from the specimen-scattered, foreground, details.
- Increases contrast for improved visibility.
Specifications
Specifications
| Type | Phase Contrast Slider |
| For Use With (Equipment) | CKX53 and IX53 Microscopes |