missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CKS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570531
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 79%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%; Zebrafish: 93%.
CKS1B protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS1B mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects a specialized role for the encoded protein. At least two transcript variants have been identified for this gene, and it appears that only one of them encodes a protein.
Specifications
| CKS1 | |
| Polyclonal | |
| Unconjugated | |
| CKS1B | |
| 2410005G18Rik; 2610005D03Rik; AA407784; CDC28 protein kinase 1; CDC28 protein kinase 1B; CDC28 protein kinase regulatory subunit 1; CDC28 protein kinase regulatory subunit 1B; CDC2-associated protein CKS1; cell division control protein CKS1; CKS1; CKS-1; Cks1b; ckshs1; cyclin-dependent kinase regulatory subunit 1; cyclin-dependent kinases regulatory subunit 1; NB4 apoptosis/differentiation related protein; PNAS-143; PNAS-16; PNAS-18; RGD1561797; Sid 1334; sid1334; Unknown (protein for MGC:142665); zgc:101026 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54124 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P61025 | |
| CKS1B | |
| synthetic peptide directed towards the middle region of human CKS1 (aa 16-65). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction