missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CKS1 Antibody (3G8), Novus Biologicals™

Product Code. 18344598 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344598 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344598 Supplier Novus Biologicals™ Supplier No. H00001163M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CKS1 Monoclonal antibody specifically detects CKS1 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKS1
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 3G8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001817
Gene Alias CDC28 protein kinase 1, CDC28 protein kinase 1B, CDC28 protein kinase regulatory subunit 1B, cell division control protein CKS1, CKS-1, CKS1CDC2-associated protein CKS1, ckshs1, cyclin-dependent kinases regulatory subunit 1, NB4 apoptosis/differentiation related protein, PNAS-143, PNAS-16, PNAS-18
Host Species Mouse
Immunogen CKS1B (NP_001817, 1 a.a. ∽ 79 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSHKQIYYSDKYDDEEFEYRHVMLPKDIAKLVPKTHLMSESEWRNLGVQQSQGWVHYMIHEPEPHILLFRRPLPKKPKK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1163
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.