missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKMT2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CKMT2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602992
|
Novus Biologicals
NBP2-92825-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644812
|
Novus Biologicals
NBP2-92825-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CKMT2 Polyclonal antibody specifically detects CKMT2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CKMT2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1160 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Basic-type mitochondrial creatine kinase, creatine kinase S-type, mitochondrial, creatine kinase, mitochondrial 2 (sarcomeric), EC 2.7.3, EC 2.7.3.2, mib-CK, Sarcomeric mitochondrial creatine kinase, S-MtCK, SMTCK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-230 of human CKMT2 (NP_001816.2). EVREQPRLFPPSADYPDLRKHNNCMAECLTPAIYAKLRNKVTPNGYTLDQCIQTGVDNPGHPFIKTVGMVAGDEESYEVFADLFDPVIKLRHNGYDPRVMKHTTDLDASKITQGQFDEHYVLSSRVRTGRSIRGLSLPPACTRAERREVENVAITALEGLKGDLAGRYYKLSEMTEQDQQRLIDDHFLFDK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title