missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKM Mouse anti-Human, Mouse, Rat, Clone: 2G3F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60177-1-IG-20UL
This item is not returnable.
View return policy
Description
Creatine kinase isoenzymes play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa. The CKM isoform, predomit in skeletal muscle and heart tissue, is a cytoplasmic enzyme involved in energy homeostasis and is an important serum marker for myocardial infarction. CKM reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in striated muscle as well as in other tissues, and as a heterodimer with a similar brain isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family.Specifications
| CKM | |
| Monoclonal | |
| 2.03 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P00564, P06732, P07310 | |
| Ckm | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG2a |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 2G3F6 | |
| Unconjugated | |
| CKM | |
| CKMM, Creatine Kinase MM, Creatine kinase M chain, Creatine kinase M type, creatine kinase, muscle, M CK | |
| Mouse | |
| Protein A | |
| RUO | |
| 1158, 12715, 24265 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction