missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKM/CKB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15891-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize both CKB and CKM due to the high homology.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis and is an important serum marker for myocardial infarction. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in striated muscle as well as in other tissues, and as a heterodimer with a similar brain isozyme in heart. The encoded protein is a member of the ATP: guanido phosphotransferase protein family.Specifications
| CKM/CKB | |
| Polyclonal | |
| Unconjugated | |
| CKB | |
| CKMM, Creatine Kinase, Creatine Kinase MM, Creatine kinase M chain, Creatine kinase M type, creatine kinase, muscle, M CK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1152, 1158, 12709, 12715, 24264, 24265 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00564, P06732, P07310, P07335, P12277, Q04447 | |
| CKB, Ckm | |
| CKM/CKB Fusion Protein Ag8678 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction