missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMTM1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total CMTM1 protein.
Chemokines are members of a superfamily of small inducible, secreted, pro-inflammatory cytokines. Members of the chemokine-like factor superfamily (CKLFSF) share similarities with both chemokine and transmembrane 4 superfamilies of signaling molecules. As a member of the CKLFSF family of proteins, CMTM1 (CKLF-like MARVEL transmembrane domain-containing protein 1), also known as CKLFSF1 (Chemokine-like factor superfamily member 1), is a 169 amino acid multi-pass membrane protein that is highly expressed in testis, therefore probably playing a role in spermatogenesis or testicular development. CMTM1 contains a CC-chemokine motif and four transmembrane segments. The gene encoding CMTM1 resides in a tight gene cluster on human chromosome 16 with CKLF, CMTM2, CMTM3 and CMTM4. There are at least sixteen isoforms of CMTM1 that are produced as a result of alternative splicing events.
Specifications
Specifications
| Antigen | CMTM1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CMTM1 |
| Gene Accession No. | Q8IZ96 |
| Gene Alias | chemokine-like factor super family 1; chemokine-like factor superfamily 1; chemokine-like factor superfamily member 1; chemokine-like factor-like protein CKLFH1; CHLFH1a; CKLF like MARVEL transmembrane domain containing 1; CKLFH; CKLFH1; CKLF-like MARVEL transmembrane domain containing 1; CKLF-like MARVEL transmembrane domain-containing protein 1; CKLFSF1; CMTM1 |
| Gene Symbols | CMTM1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?