missing translation for 'onlineSavingsMsg'
Learn More

CKIP-1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18401434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401434 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401434 Supplier Novus Biologicals Supplier No. NBP176238MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CKIP-1 Polyclonal antibody specifically detects CKIP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKIP-1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 2810052M02Rik, Casein kinase 2-interacting protein 1, C-Jun-binding protein, CK2 interacting protein 1; HQ0024c protein, CK2-interacting protein 1, CKIP1, CKIP-1PH domain-containing family O member 1, JBP, OC120, Osteoclast maturation-associated gene 120 protein, pleckstrin homology domain containing, family O member 1, pleckstrin homology domain-containing family O member 1
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human CKIP1. The immunogen is located within the last 50 amino acids of CKIP1. Amino Acid Squence: TPDSHLRQTTPHSQYKKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51177
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.