missing translation for 'onlineSavingsMsg'
Learn More

CKIP-1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18425493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425493 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425493 Supplier Novus Biologicals Supplier No. NBP176238UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CKIP-1 Polyclonal antibody specifically detects CKIP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKIP-1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias 2810052M02Rik, Casein kinase 2-interacting protein 1, C-Jun-binding protein, CK2 interacting protein 1; HQ0024c protein, CK2-interacting protein 1, CKIP1, CKIP-1PH domain-containing family O member 1, JBP, OC120, Osteoclast maturation-associated gene 120 protein, pleckstrin homology domain containing, family O member 1, pleckstrin homology domain-containing family O member 1
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human CKIP1. The immunogen is located within the last 50 amino acids of CKIP1. Amino Acid Squence: TPDSHLRQTTPHSQYKKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51177
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.