missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKII alpha prime polypeptide Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37929-20ul
This item is not returnable.
View return policy
Description
CKII alpha prime polypeptide Polyclonal antibody specifically detects CKII alpha prime polypeptide in Human samples. It is validated for ELISA,Western Blot
Specifications
| CKII alpha prime polypeptide | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| casein kinase 2, alpha prime polypeptide, casein kinase II subunit alpha', CK II alpha', CK2A2, CSNK2A1, EC 2.7.11, EC 2.7.11.1, FLJ43934 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human CKII alpha prime polypeptide (NP_001887.1).,, Sequence:, MPGPAAGSRARVYAEVNSLRSREYWDYEAHVPSWGNQDDYQLVRKLGRGKYSEVFEAINITNNERVVVKILKPVKKKKIKREVKILENLRGGTNIIKLIDTVKDPVSKTPALVFEYINNTDFKQLYQILTDFDIRFYMYELLKALDYCHSKGIMHRDVKPHNVMIDHQQKKLRLIDWGLAEFYHPAQEYNVRVASRYFKGPELLVDYQMYDYSLDMWSLGCMLASMIFRREPFFHGQDNYDQLVRIAKVLGTEELYGYLKKYHIDLDPHFNDILGQHSRKRWENFIHSENRHLVSPEALDLLDKLLRYDHQQRLTAKEAMEHPYFYPVVKEQSQPCADNAVLSSGLTAAR | |
| 20 μL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 1459 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction