missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKII alpha prime polypeptide Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99686CL1
This item is not returnable.
View return policy
Description
CKII alpha prime polypeptide Polyclonal antibody specifically detects CKII alpha prime polypeptide in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CKII alpha prime polypeptide | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| casein kinase 2, alpha prime polypeptide, casein kinase II subunit alpha', CK II alpha', CK2A2, CSNK2A1, EC 2.7.11, EC 2.7.11.1, FLJ43934 | |
| Produced in rabbits immunized with purified, recombinant Human CKII alpha prime polypeptide (Accession#: NP_001887.1; Met1-Arg350) | |
| 0.1 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 1459 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction