missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKI gamma 3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92102-0.02ml
This item is not returnable.
View return policy
Description
CKI gamma 3 Polyclonal antibody specifically detects CKI gamma 3 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| CKI gamma 3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| casein kinase 1, gamma 3, casein kinase I isoform gamma-3, CKI-gamma 3, EC 2.7.11, EC 2.7.11.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 304-423 of human CSNK1G3 (NP_001026982.1). LRKLFTDLFDRKGYMFDYEYDWIGKQLPTPVGAVQQDPALSSNREAHQHRDKMQQSKNQVVSSTNGELNTDDPTAGRSNAPITAPTEVEVMDETNCQKVLNMWCCCFFKRRKRKTIQRHK | |
| 0.02 mL | |
| Protein Kinase | |
| 1456 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction