missing translation for 'onlineSavingsMsg'
Learn More

CKAP2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18418433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418433 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418433 Supplier Novus Biologicals Supplier No. NBP121391JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CKAP2 Polyclonal antibody specifically detects CKAP2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP2
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CTCL tumor antigen se20-10, cytoskeleton associated protein 2, cytoskeleton-associated protein 2, LB1DKFZp686L1238, se20-10, TMAPFLJ10749, Tumor- and microtubule-associated protein, tumor-associated microtubule-associated protein
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 633 and 683 of human cytoskeleton associated protein 2 using the numbering given in entry NP_001091995.1 (GeneID 26586).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26586
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.