missing translation for 'onlineSavingsMsg'
Learn More

CKAP2 Antibody (3H1), Novus Biologicals™

Product Code. 18338589 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338589 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338589 Supplier Novus Biologicals Supplier No. H00026586M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CKAP2 Monoclonal antibody specifically detects CKAP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3H1
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH10901
Gene Alias CTCL tumor antigen se20-10, cytoskeleton associated protein 2, cytoskeleton-associated protein 2, LB1DKFZp686L1238, se20-10, TMAPFLJ10749, Tumor- and microtubule-associated protein, tumor-associated microtubule-associated protein
Host Species Mouse
Immunogen CKAP2 (AAH10901.1, 1 a.a. ~ 161 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEKSEPVDQRRHTAGKAIVDSRSAQPKETSEERKARLSEWKAGKGRVLKRPPNSVVTQHEPAGQNEKPVGSFWTTMAEEDEQRLFTEKVNNTFSECLNLINEGCPKEDILVTLNDLIKNIPDAKKLVKYWICLALIEPITSPIENIIAIYEKAILAGAQVR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26586
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.