missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CKAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587159
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
CKAP1 binds to alpha-tubulin folding intermediates after their interaction with cytosolic chaperonin in the pathway leading from newly synthesized tubulin to properly folded heterodimer. Involved in regulation of tubulin heterodimer dissociation. May function as a negative regulator of axonal growth.
Specifications
| CKAP1 | |
| Polyclonal | |
| Unconjugated | |
| TBCB | |
| 2410007D12Rik; AU041393; CG22; Ckap1; CKAPI; cytoskeleton associated protein 1; cytoskeleton-associated protein 1; Cytoskeleton-associated protein CKAPI; liver cancer related protein; TBCB; tubulin folding cofactor B; tubulin-folding cofactor B; Tubulin-specific chaperone B; ZH14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1155 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99426 | |
| TBCB | |
| Recombinant Protein of human TBCB. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction