missing translation for 'onlineSavingsMsg'
Learn More

CKAP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18382132 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382132 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382132 Supplier Novus Biologicals Supplier No. NBP300264AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CKAP1 Polyclonal specifically detects CKAP1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CG22Tubulin-specific chaperone B, CKAP1MGC14625, CKAPI, cytoskeleton associated protein 1, Cytoskeleton-associated protein 1Cytoskeleton-associated protein CKAPI, tubulin folding cofactor B, tubulin-folding cofactor B
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CKAP1 (Q99426; Met1-Ile244).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1155
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.