missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK2 beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA582226
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: PHYRQALDMI LDLEPDEELE DNPNQSDLIE QAAEMLYGLI HARYILTNRG IAQMLEKYQQ GDFGYCPRVY CENQPMLPIG LSDIPGEAMV KLYCPKCMDV YTPKSSRHHH TDGAYFGTGF PHMLFMVHPE YRPKRPANQF VPRLY Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus.
Spezifikation
| CK2 beta | |
| Polyclonal | |
| Unconjugated | |
| Csnk2b | |
| alternative name: G5a, phosvitin; casein kinase 2 beta; casein kinase 2, beta polypeptide; casein kinase 2, beta subunit; casein kinase II b subunit splicing isoform 132; casein kinase II b subunit splicing isoform 247; casein kinase II b subunit splicing isoform 318; casein kinase II b subunit splicing isoform 660; casein kinase II b subunit splicing isoform 806; Casein kinase II beta subunit; casein kinase II subunit beta; chimera CSNK2B-LY6G5B splicing isoform 1072; chimera CSNK2B-LY6G5B splicing isoform 1103; chimera Csnk2b-Ly6g5b splicing isoform 2050; chimera Csnk2b-Ly6g5b splicing isoform 2275; chimera Csnk2b-Ly6g5b splicing isoform 2531; chimera CSNK2B-LY6G5B splicing isoform 532; chimera CSNK2B-LY6G5B splicing isoform 560; chimera CSNK2B-LY6G5B splicing isoform 562; chimera Csnk2b-Ly6g5b splicing isoform 901; CK II beta; Ck2; CK2B; CK2N; CSK2B; Csnk2b; Csnk2b splicing isoform 280; Csnk2b splicing isoform 662; Csnk2b-Ly6g5b; CSNK2B-LY6G5B-1072; CSNK2B-LY6G5B-1103; CSNK2B-LY6G5B-532; CSNK2B-LY6G5B-560; CSNK2B-LY6G5B-562; DADB-127H9.2; G5A; MGC138222; MGC138224; Phosvitin; protein G5a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1460 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P67870 | |
| Csnk2b | |
| Recombinant protein corresponding to Human CSNK2B. Recombinant protein control fragment (Product #RP-101869). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur