missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK2 alpha-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595701
This item is not returnable.
View return policy
Description
Immunogen sequence: LGTEELYGYL KKYHIDLDPH FNDILGQHSR KRWENFIHSE NRHLVSPEAL DLLDKLLRYD HQQRLTAKEA MEHPYFYPVV KEQSQPCADN AVLSSGLTAA R Positive Samples: K-562, 293T, HeLa, Mouse testis, Mouse brain.
Casein kinase II is a protein serine/threonine kinase that is thought to be present in all eukarotic cells, implying that it has fundamental cellular functions. The holoenzyme is a tetramer containing 2 alpha or alpha-prime subunits (or one of each) and 2 beta subunits. The function of beta subunits is unknown and the alpha subunit is the catalytic unit. CK2 functions as a tetrameric complex consisting of two regulatory beta-subunits and two catalytic units (alpha and alpha') in a homomeric or heteromeric conformation. Whilst the alpha- and alpha'-subunits are catalytically identical, proteins that regulate CK2, such as cdc2 and Hsp90, preferentially bind to the alpha and not the alpha'-subunit. CK2 can phosphorylate a number of key intracellular signaling proteins implicated in tumor suppression (p53 and PTEN) and tumorigenesis (myc, jun, NF-kappaB). CK2 is also thought to influence Wnt signaling via beta-catenin phosphorylation and the PI 3-K signaling pathway via the phosphorylation of Akt.
Specifications
| CK2 alpha-2 | |
| Polyclonal | |
| Unconjugated | |
| CSNK2A2 | |
| 1110035J23Rik; C77789; casein kinase 2 alpha; casein kinase 2 alpha'; casein kinase 2 alpha 2; casein kinase 2, alpha prime polypeptide; casein kinase II subunit alpha'; casein kinase II, alpha 2, polypeptide; CK II alpha'; CK2; CK2A2; CK2alpha'; CSNK2A1; CSNK2A2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13000, 1459, 307641 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.68 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O54833, P19784 | |
| CSNK2A2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido