missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK1 gamma-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588914
This item is not returnable.
View return policy
Description
Immunogen sequence: LRKLFTDLFD RKGYMFDYEY DWIGKQLPTP VGAVQQDPAL SSNREAHQHR DKMQQSKNQV VSSTNGELNT DDPTAGRSNA PITAPTEVEV MDETNCQKVL NMWCCCFFKR RKRKTIQRHK; Positive Samples: Mouse eye, Mouse brain, Rat heart; Cellular Location: Cytoplasm.
The casein kinase I (CKI) gene family encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. In eukaryotic cells, CKI is in volved in regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
Specifications
| CK1 gamma-3 | |
| Polyclonal | |
| Unconjugated | |
| CSNK1G3 | |
| 3300002K07Rik; C330049O21Rik; casein kinase 1 gamma 3; casein kinase 1, gamma 3; casein kinase I isoform gamma-3; CKI-gamma 3; CSNK1G3; CSNK1G3L | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64823, 70425 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.68 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q62763, Q8C4X2 | |
| CSNK1G3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 304-423 of human CSNK1G3 (NP_0010269821). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction