missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CK1 gamma-2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583249

Product Code. 16380205

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FDKKGGKGET EEGRRMSKAG GGRSSHGIRS S Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 81%.

Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CK1 gamma-2
Polyclonal
Unconjugated
Csnk1g2
2810429I12Rik; AI463719; casein kinase 1 gamma 2; casein kinase 1 gamma 2 isoform; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 2; casein kinase I isoform gamma-2; CK Igamma2; CK1G2; CKI gamma 2; CKI gamma2; CKI-gamma 2; CKIgamma2; CK-Igamma2; CKI-gamma2; CKI-gamma-2; CKIgamma-2; Csnk1g2
Rabbit
Antigen affinity chromatography
RUO
1455
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P78368
Csnk1g2
Recombinant protein corresponding to Human CSNK1G2. Recombinant protein control fragment (Product #RP-97246).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.