missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK1 gamma-1 Monoclonal Antibody (OTI1C5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526787
This item is not returnable.
View return policy
Description
CK1 gamma-1 Monoclonal Antibody for Western Blot, IHC (P)
Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha' beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.
Specifications
| CK1 gamma-1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9HCP0 | |
| Csnk1g1 | |
| Human recombinant protein fragment corresponding to amino acids 163-422 of CSNK1G1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C5 | |
| Unconjugated | |
| Csnk1g1 | |
| 9130020E21Rik; casein kinase 1 gamma 1; casein kinase 1, gamma 1; casein kinase I isoform gamma-1; casein kinase I, gamma 1; CK I gamma1; CK1G1; CK1gamma1; CKI gamma 1; CKI gamma1; CK-I gamma1; CKI- gamma1; CKI gamma-1; CKI-gamma 1; CKI-gamma-1; CSNK1G1; KC1G1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 53944 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction