missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK1 delta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597636
This item is not returnable.
View return policy
Description
CK1 delta Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA, IP
CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
Specifications
| CK1 delta | |
| Polyclonal | |
| Unconjugated | |
| CSNK1D | |
| 1200006A05Rik; AA409348; ASPS; casein kinase 1; casein kinase 1 delta; casein kinase 1, delta; casein kinase I isoform delta; CK1d; CK1delta; CKId; CKIdelta; CKI-delta; CSNK1D; D930010H05Rik; FASPS2; HCKID; HGNC:2452; Tau-protein kinase CSNK1D | |
| Rabbit | |
| Protein G | |
| RUO | |
| 104318, 1453, 64462 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P48730, Q06486, Q9DC28 | |
| CSNK1D | |
| Recombinant Human Casein kinase I isoform delta protein (152-398AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction