missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CK068 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17151184 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17151184 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17151184 Supplier Invitrogen™ Supplier No. PA5113324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CK068 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CK068
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene C11orf68
Gene Accession No. Q9H3H3
Gene Alias basophilic leukemia expressed protein BLES03; basophilic leukemia-expressed protein Bles03; BLES03; C11orf68; chromosome 11 open reading frame 68; P5326; protein p5326; UPF0696 protein C11orf68
Gene Symbols C11orf68
Host Species Rabbit
Immunogen Recombinant Human UPF0696 protein C11orf68 protein (1-251AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83638
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.