missing translation for 'onlineSavingsMsg'
Learn More

CK-666, MedChemExpress

Product Code. p-200100429
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489737

Brand: Medchem Express HY16926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CK-666 is a cell-permeable actin-related protein Arp2/3 complex inhibitor (IC50=12 μM). CK-666 binds to Arp2/3 complex, stabilizes the inactive state of the complex, blocking movement of the Arp2 and Arp3 subunits into the activated filament-like (short pitch) conformation.

CK-666 is a cell-permeable actin-related protein Arp2/3 complex inhibitor (IC50=12 μM). CK-666 binds to Arp2/3 complex, stabilizes the inactive state of the complex, blocking movement of the Arp2 and Arp3 subunits into the activated filament-like (short pitch) conformation.

Specifications

Chemical Name or Material CK-666
CAS 442633-00-3
Color White
Purity Grade Notes Research
Molecular Formula C18H17FN2O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (421.81 mM; Need ultrasonic)
SMILES O=C(NCCC1=C(C)NC2=C1C=CC=C2)C3=CC=CC=C3F
Molecular Weight (g/mol) 296.34
Formula Weight 296.34
Percent Purity 99.79%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.