missing translation for 'onlineSavingsMsg'
Learn More

CK-666, MedChemExpress

Product Code. 16489737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489737 5 mg 5mg
16417218 10 mg 10mg
16413237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489737 Supplier Medchem Express Supplier No. HY16926
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CK-666 is a cell-permeable actin-related protein Arp2/3 complex inhibitor (IC50=12 μM). CK-666 binds to Arp2/3 complex, stabilizes the inactive state of the complex, blocking movement of the Arp2 and Arp3 subunits into the activated filament-like (short pitch) conformation.

CK-666 is a cell-permeable actin-related protein Arp2/3 complex inhibitor (IC50=12 μM). CK-666 binds to Arp2/3 complex, stabilizes the inactive state of the complex, blocking movement of the Arp2 and Arp3 subunits into the activated filament-like (short pitch) conformation.

Specifications

Chemical Name or Material CK-666
CAS 442633-00-3
Color White
Purity Grade Notes Research
Molecular Formula C18H17FN2O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (421.81 mM; Need ultrasonic)
SMILES O=C(NCCC1=C(C)NC2=C1C=CC=C2)C3=CC=CC=C3F
Molecular Weight (g/mol) 296.34
Formula Weight 296.34
Percent Purity 99.79%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.