missing translation for 'onlineSavingsMsg'
Learn More

CK-636, MedChemExpress

Product Code. 16442967
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16442967

Brand: Medchem Express HY15892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CK-636 is a cell permeable inhibitor of Arp2/3 complex, that could inhibit actin polymerization, with IC50 values of 4 μM, 24 μM and 32 μM for human, fission yeast and bovine, respectively.

CK-636 is a cell permeable inhibitor of Arp2/3 complex, that could inhibit actin polymerization, with IC50 values of 4 μM, 24 μM and 32 μM for human, fission yeast and bovine, respectively.

Specifications

Chemical Name or Material CK-636
Physical Form Solid
CAS 442632-72-6
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H16N2OS
For Use With (Application) Cancer-programmed cell death
Synonym CK-0944636
Solubility Information DMSO : ≥ 49 mg/mL (172.30 mM)
SMILES O=C(C1=CC=CS1)NCCC2=C(C)NC3=C2C=CC=C3
Molecular Weight (g/mol) 284.38
Formula Weight 284.38
Percent Purity 98.43%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.