missing translation for 'onlineSavingsMsg'
Learn More

CIZ1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-74623

Product Code. 18017995

  • £464.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CIZ1 Polyclonal specifically detects CIZ1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CIZ1
Polyclonal
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
CDKN1A interacting zinc finger protein 1, CDKN1A-interacting zinc finger protein 1, cip1-interacting zinc finger protein, LSFR1ZNF356Nuclear protein NP94, NP94, Zinc finger protein 356
Rabbit
Affinity Purified
RUO
Primary
Human
IgG
Western Blot, Immunoprecipitation
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
NP_036259.2
CIZ1
The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human CDKN1A interacting zinc finger protein 1 using the numbering given in entry NP_036259.2 (GeneID 25792).
0.1 mL
Cancer
25792
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only