missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIZ1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74624AF532
This item is not returnable.
View return policy
Description
CIZ1 Polyclonal antibody specifically detects CIZ1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CIZ1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CDKN1A interacting zinc finger protein 1, CDKN1A-interacting zinc finger protein 1, cip1-interacting zinc finger protein, LSFR1ZNF356Nuclear protein NP94, NP94, Zinc finger protein 356 | |
| The immunogen recognized by this antibody maps to a region between residue 848 and 898 of human CDKN1A interacting zinc finger protein 1 using the numbering given in entry NP_036259.2 (GeneID 25792). | |
| 0.1 mL | |
| Cancer | |
| 25792 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction