missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citrulline Antibody (6C2.1), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59365C
This item is not returnable.
View return policy
Description
Citrulline Monoclonal specifically detects Citrulline in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Citrulline | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 6C2.1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| ACPA, Citrulline, L-Citrulline | |
| Synthetic L-Citrulline conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 100 μL | |
| Specific for citrulline residues on modified proteins. Tested using citrulline conjugated Bovine Serum Albumin (BSA). | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction