missing translation for 'onlineSavingsMsg'
Learn More

Citronellyl acetate, 96%

Product Code. 11448707
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448707 100 g 100g
11438707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448707 Supplier Thermo Scientific Alfa Aesar Supplier No. A19872.22
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 150-84-5 | C12H22O2 | 198.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-84-5
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00015039
InChI Key JOZKFWLRHCDGJA-UHFFFAOYSA-N
Synonym citronellyl acetate, citronellol acetate, 3,7-dimethyl-6-octen-1-yl acetate, 3,7-dimethyloct-6-en-1-yl acetate, 1-acetoxy-3,7-dimethyloct-6-ene, acetic acid, citronellyl ester, natural rhodinol, acetylated, citronellyl ethanoate, b-citronellyl acetate, 6-octen-1-ol, 3,7-dimethyl-, acetate
PubChem CID 9017
ChEBI CHEBI:70478
IUPAC Name 3,7-dimethyloct-6-enyl acetate
SMILES CC(CCC=C(C)C)CCOC(=O)C

Specifications

Density 0.89
Boiling Point 240°C
Flash Point 98°C (208°F)
Refractive Index 1.4434
Quantity 100 g
Formula Weight 198.3
Percent Purity 96%
Chemical Name or Material Citronellyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.