missing translation for 'onlineSavingsMsg'
Learn More

Citron Kinase Antibody, DyLight 488, Novus Biologicals™

Product Code. 18405333 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405333 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405333 Supplier Novus Biologicals Supplier No. NBP128740G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Citron Kinase Polyclonal antibody specifically detects Citron Kinase in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Citron Kinase
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CITK, citron (rho-interacting, serine/threonine kinase 21), Citron Rho-Interacting Kinase, Citron Rho-Interacting Serine/Threonine Kinase, CRIK, KIAA0949, MCPH17, Rho-interacting Serine/Threonine kinase 21, serine/threonine kinase 21, Serine/threonine-protein kinase 21, STK21
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1925 and 1975 of human citron (rho-interacting, serine/threonine kinase 21) using the numbering given in entry NP_009105.1 (GeneID 11113).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 11113
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.