missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Absorbance | 0.02 max. (0.1 M in water) at 260nm |
Melting Point | >300.0°C |
Color | White |
pH | 7 to 9 (5% aq. soln.) |
Loss on Drying | 12% to 13% |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.998 |
Packaging | Plastic bottle |
Quantity | 250 g |
Merck Index | 14, 8602 |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'