missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and Protease free

Product Code. p-3757409 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10590994

Brand: Thermo Scientific Acros 327162500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114456-61-0 | C6H5Na3O7 | 258.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114456-61-0
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00150032
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Synonym trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
PubChem CID 21868242
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Absorbance 0.02 max. (0.1 M in water) at 260nm
Melting Point >300.0°C
Color White
pH 7 to 9 (5% aq. soln.)
Loss on Drying 12% to 13%
Infrared Spectrum Authentic
Assay Percent Range 0.998
Packaging Plastic bottle
Quantity 250 g
Merck Index 14, 8602
Solubility Information Solubility in water: soluble.
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
Formula Weight 258.08
Percent Purity 0.998
Grade Molecular Biology
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Citric acid, trisodium salt hydrate, For molecular biology
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.