missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trisodium salt dihydrate, for HPLC, Thermo Scientific™

Product Code. 11905071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
500 g, Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11905071 500 g, Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11905071 Supplier Thermo Scientific Acros Supplier No. 385925000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4-3-6132
Melting Point >300.0°C
Molecular Formula C6H5Na3O7·2H2O
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2 H2O
Merck Index 15, 8737
Solubility Information Solubility aq. soln.: 720 g/L (25°C). Other solubilities: insoluble in alcohol
Molecular Weight (g/mol) 294.09
Formula Weight 294.09
Grade HPLC
Quantity 500 g
Packaging Plastic bottle
Chemical Name or Material Citric acid, trisodium salt dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.