missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trilithium salt tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Product Code. p-93297 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
500g
5g
This item is not returnable. View return policy

Product Code. 10511841

Brand: Thermo Scientific Acros 199850050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-58-6
MDL Number MFCD00150030
InChI Key HXGWMCJZLNWEBC-UHFFFAOYSA-K
Synonym lithium citrate tetrahydrate, unii-5z6e9k79yv, trilithium citrate tetrahydrate, lithium citrate usp, citric acid trilithium salt tetrahydrate, tri-lithium citrate tetrahydrate, lithium citrate tribasic tetrahydrate, lithonate tn, 1,2,3-propanetricarboxylic acid, 2-hydroxy-trilithium salt tetrahydrate
PubChem CID 2724118
ChEBI CHEBI:64754
IUPAC Name trilithium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
SMILES [Li+].[Li+].[Li+].C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O

Specifications

Melting Point 112.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H13Li3O11
Linear Formula LiO2CCH2C(OH)(CO2Li)CH2CO2Li·4H2O
Quantity 5 g
Beilstein 03, I, 196
Merck Index 15, 5586
Solubility Information Solubility in water: 745g/L water (25°C). Other solubilities: slightly soluble in alcohol
Molecular Weight (g/mol) 281.979
Formula Weight 281.97
Percent Purity 98%
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Citric acid, trilithium salt tetrahydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.