missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Solution, 50% by weight, Biotech Reagent, J.T. Baker™

Product Code. 15557824 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
19 L
Packaging:
Poly Bottle
Unit Size:
19 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15557824 19 L Poly Bottle 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557824 Supplier Avantor J.T.Baker Supplier No. 034107
  • ONLINE EXCLUSIVE PRICE
    £1384.50
    £1044.66 / 19 L
    Save £339.84
    24% Off
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Specifications

CAS 7664-41-7
Molecular Formula H3N
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES N
IUPAC Name ammonia
Molecular Weight (g/mol) 17.03
Formula Weight 192.12
Percent Purity 50%
Grade Biotech Reagent
Quantity 19 L
Packaging Poly Bottle
Specific Gravity 1L = 1.24kg
Chemical Name or Material Citric Acid Solution
Show More Show Less
compliance-icons
Product Identifier
  • Citric Acid Solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.