missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, TRC

Product Code. 30133597
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133597 50 g 50g
30011406 10 g 10g
30011048 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133597 Supplier Toronto Research Chemicals Supplier No. TRCC52105850G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6 H8 O7 . H2 O
Molecular Weight (g/mol) 210.14
Synonym 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, monohydrate (9CI), Citric acid, monohydrate (8CI), 2-Hydroxy-1,2,3-propanetricarboxylic acid hydrate (1:1), 2-Hydroxy-1,2,3-propanetricarboxylic acid monohydrate, Citronsyra Mono E 33-8-80MLT
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES O.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2
Formula Weight 210.0376
Percent Purity >95%
Quantity 50 g
Chemical Name or Material Citric acid monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.