missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™

Product Code. 10665102 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.5kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665102 5 kg Plastic keg 5kg
10345410 500 g Plastic powder jar 500g
10655102 1 kg Plastic powder jar 1kg
10796281 2.5 kg Plastic powder jar 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10665102 Supplier Fisher Chemical Supplier No. C/6200/65
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C6H8O7.H2O, CAS Number-5949-29-1, 5kg, White, 210.15g/mol, 135 deg.C, 149972, Plastic keg, 1.8, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
MDL Number 149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 1.8
Melting Point 135°C
Color White
Formula Weight 210.15g/mol
Quantity 5 kg
Packaging Plastic keg
Physical Form Solid
compliance-icons
Product Identifier
  • Citric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid monohydrate, Citric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.