missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£180.00
Chemical Identifiers
CAS | 5949-29-1 |
---|---|
Molecular Formula | C6H10O8 |
Molecular Weight (g/mol) | 210.138 |
InChI Key | YASYEJJMZJALEJ-UHFFFAOYSA-N |
Synonym | citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp |
PubChem CID | 22230 |
ChEBI | CHEBI:31404 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
Chemical Identifiers
5949-29-1 | |
210.138 | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
CHEBI:31404 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
C6H10O8 | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
22230 | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
Specifications
5949-29-1 | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O | |
210.138 | |
CHEBI:31404 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 1.54kg |
C6H10O8 | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate | |
22230 | |
210.14 | |
Granular | |
Citric Acid |