missing translation for 'onlineSavingsMsg'
Learn More

Citric acid diammonium salt, 99+%

Product Code. 11397528
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397528 100 g 100g
11317538 500 g 500g
11307538 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397528 Supplier Thermo Scientific Alfa Aesar Supplier No. 033221.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol

Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3012-65-5
Molecular Formula C6H14N2O7
Molecular Weight (g/mol) 226.19
MDL Number MFCD00013068
InChI Key YXVFQADLFFNVDS-UHFFFAOYSA-N
Synonym ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
PubChem CID 13710713
ChEBI CHEBI:63076
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O

Specifications

Color White
Density 1.48 g/mL
Odor Odorless
Linear Formula HOC(CO2H)(CH2CO2NH4)2
Quantity 100 g
Merck Index 14512
Solubility Information Soluble in water. Slightiy soluble in alcohol.
Formula Weight 226.19
Percent Purity ≥99%
Physical Form Granules
Chemical Name or Material Citric acid diammonium salt
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.