missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43 Rat anti-Mouse, BB700, Clone: S7, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566508
This item is not returnable.
View return policy
Description
CD43 Monoclonal antibody specifically detects CD43 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD43 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| CD43 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| S7 | |
| Brilliant Blue 700 (BB700) | |
| 0.1 mg | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction