missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Citric Acid Anhydrous
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 77-92-9
- Molecular Formula: C6H8O7
- Molecular Weight: 192.1
Specifications
Specifications
| Chemical Name or Material | Citric Acid Anhydrous |
| CAS | 77-92-9 |
| Molecular Formula | C6H8O7 |
| Quantity | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction